1,2-Dichloro-4-iodobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Dichloroiodobenzene

HC-2592-5G HC-2592-25G HC-2592-100G
COBBHC-2592-5GEA 0 GBP
COBBHC-2592-5G COBBHC-2592-25G COBBHC-2592-100G
1,2-Dichloro-4-iodobenzene
1,2-Dichloro-4-iodobenzene
Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
MDL Number: MFCD00019014
CAS Number: 20555-91-3
EINECS: 243-874-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR