1,2-Dichloro-3-iodobenzene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
HC-2711-25G
HC-2711-5G
HC-2711-100G
COBBHC-2711-25GEA
0
GBP
COBBHC-2711-25G
COBBHC-2711-5G
COBBHC-2711-100G
1,2-Dichloro-3-iodobenzene
1,2-Dichloro-3-iodobenzene
Formula:
C₆H₃Cl₂I MW: 272.9 g/mol Boiling Pt: 242 °C (1013 hPa) Melting Pt: 35 °C |
MDL Number:
MFCD00001034 CAS Number: 2401-21-0 EINECS: 219-276-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...