3,4-Dichloro-2,6-dinitrotoluene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1424-1G
AN-1424-5G
COBBAN-1424-1GEA
0
GBP
COBBAN-1424-1G
COBBAN-1424-5G
3,4-Dichloro-2,6-dinitrotoluene
3,4-Dichloro-2,6-dinitrotoluene
Formula:
C₇H₄Cl₂N₂O₄ MW: 251.03 g/mol |
MDL Number:
MFCD11855827 CAS Number: 53278-85-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...