1,2-Dibromo-4-nitrobenzene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1296-25G
AN-1296-1G
AN-1296-5G
COBBAN-1296-25GEA
0
GBP
COBBAN-1296-25G
COBBAN-1296-1G
COBBAN-1296-5G
1,2-Dibromo-4-nitrobenzene
1,2-Dibromo-4-nitrobenzene
Formula:
C₆H₃Br₂NO₂ MW: 280.9 g/mol |
MDL Number:
MFCD00055373 CAS Number: 5411-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...