4'-tert-Butylacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
p-tert-Butylacetophenone
002522-5G
002522-25G
FLUO002522-5GEA
0
GBP
FLUO002522-5G
FLUO002522-25G
4'-tert-Butylacetophenone
4'-tert-Butylacetophenone
Formula:
C₁₂H₁₆O MW: 176.26 g/mol Boiling Pt: 71…72 °C (0.09 mmHg) Melting Pt: 18 °C Density: 0,9705 g/cm³ (0 °C) |
MDL Number:
MFCD00017256 CAS Number: 943-27-1 EINECS: 213-399-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...