Tris(perfluorobutyl)amine

Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Perfluorotri-n-butylaminePerfluorotributylamineTris(nonafluorobutyl)aminePFTBAFC-43HeptacosafluorotributylamineHeptacosafluorotri-n-butylamine

FLUO008740-5GEA 0 GBP
FLUO008740-10X5G FLUO008740-5G FLUO008740-5X5G
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine
Formula: C₁₂F₂₇N
MW: 671.1 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –55 °C
Density: 1,883 g/cm³ (20 °C)
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR