1-(2-Bromoethoxy)-4-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1134-5G AN-1134-25G
COBBAN-1134-5GEA 0 GBP
COBBAN-1134-5G COBBAN-1134-25G
1-(2-Bromoethoxy)-4-nitrobenzene
1-(2-Bromoethoxy)-4-nitrobenzene
Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
MDL Number: MFCD00031363
CAS Number: 13288-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR