Tyramine

Supplier: LGC Standards PROMOCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Hydroxyphenethylamine4-(2-Aminoethyl)phenol

PROOMM0063.06EA 991 GBP
PROOMM0063.06
Tyramine
Tyramine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 164 °C
MDL Number: MFCD00008193
CAS Number: 51-67-2
EINECS: 200-115-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR