(4-Hydroxyphenyl)acetic acid ethyl ester

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl (4-hydroxyphenyl)acetate, Ethyl 2-(4-hydroxyphenyl)acetate

E2180.10 E2180.50
XXXXE2180.10EA 0 GBP
XXXXE2180.10 XXXXE2180.50
(4-Hydroxyphenyl)acetic acid ethyl ester
(4-Hydroxyphenyl)acetic acid ethyl ester
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00016491
CAS Number: 17138-28-2
EINECS: 241-197-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR