Sodium dihydrogen phosphate dihydrate
Supplier: MARKET SOURCE PART PROCESS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Sodium phosphate monobasic dihydrate
, Sodium dihydrogen orthophosphate dihydrate
, Phosphoric acid (sodium hydrate)
MSPPS/3760/53-500GEA
0
GBP
MSPPS/3760/53-500G
Sodium dihydrogen phosphate dihydrate
Sodium dihydrogen phosphate dihydrate
Formula:
NaH₂PO₄·2H₂O MW: 156.01 g/mol Boiling Pt: ∼650 °C (1013 hPa) Melting Pt: 60 °C Density: 1.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149209 CAS Number: 13472-35-0 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...