Formaldehyde 37% in aqueous solution

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

EMR140001 EMR140250 EMR140500
EURLEMR140001EA 0 GBP
EURLEMR140001 EURLEMR140250 EURLEMR140500
Formaldehyde 37% in aqueous solution
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR