Tubing, silicone, for perimatic GP peristaltic pump dispenser
Supplier: Jencons
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JENC256-028EA 105 GBP
JENC256-028 JENC256-029 JENC256-027
Tubing, silicone, for perimatic GP peristaltic pump dispenser
Silicone tubing assemblies.
  • Very good chemical resistance
  • Very good temperature resistance
Caution: Tubing assembly consists of tubing, glass dispense nozzle, glass 'j' piece and glass sinker