Poly-D-lysine/laminin culture dishes, Corning® BioCoat™
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Corning® BioCoat®
734-0140EA 283 GBP
734-0140
Poly-D-lysine/laminin culture dishes, Corning® BioCoat™
Culture Dishes Cell Culture Dishes
PS coated with a uniform application of poly-D-lysine and laminin. Promotes neuronal cell attachment and differentiation for a variety of applications.
Promotes neuronal cell attachment and differentiation for a variety of applications.