Spray gun assembly, chromatography
Supplier: Dixon Science
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
560-0010EA 727 GBP
560-0010
Spray gun assembly, chromatography
Atomisers
The spray gun assembly consists of a 250 ml conical glass flask (FE 250/3; 201-8419), an all-glass fine spray head (560-0009) with a 24/29 standard glass joint and a blowing ball (612-9953). The spray head features an instantaneous pressure relief vent to affect a sharp cut-off at the end of spraying.