Biopsy insert pads
Supplier: Epredia
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
720-0368EA 0 GBP
720-0368
Biopsy insert pads
Biopsy Pads
Polyester urethane foam. Biopsy material is placed between two sheets in processing cassette, and can then be processed in the normal way.
  • Foam pads cut to the size of a processing cassette chamber
Dimensions: 26×31×2 mm