VWR® Hichrom Pliers, for Stainless Steel Tubing
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Avantor® Hichrom
554-7744EA 63.7 GBP
554-7744
VWR® Hichrom Pliers, for Stainless Steel Tubing
Pliers
These pliers are designed for cutting 1/16" stainless steel tubing and are especially suitable for tubing of 0,010" and 0,020" ID.
  • Leaves smooth and uniform cuts
  • De-burring may be necessary to achieve an optimum finish