Aluminium foils
Supplier: Terinex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
291-0022EA 75.6 GBP
291-0022
Aluminium foils
Aluminium Foils
Rolls of light but strong aluminium foil. Excellent barrier against vapours, light, odours and bacteria.
  • Hygienic, recyclable
  • Non toxic and imparts no taste or odour
Delivery information: Rolls supplied in a dispenser box.