Sodium diphenylamine-4-sulphonate
Supplier: Molekula
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Sodium N-phenylsulphanilate
, Sodium (4-anilinobenzenesulphonate)
, Sodium Diphenylaminesulphonate
, Diphenylamine-4-sulphonic acid, sodium salt
MOLE34781230-5GEA
0
GBP
MOLE34781230-5G
Sodium diphenylamine-4-sulphonate
Sodium diphenylamine-4-sulphonate
Formula:
C₁₂H₁₀NNaO₃S MW: 271.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036468 CAS Number: 6152-67-6 EINECS: 228-165-6 Merck Index: 13,07397 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...