Xylenol orange tetrasodium salt

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Xylenol orange water soluble2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)

MOLEM78925342EA 0 GBP
MOLEM78925342
Xylenol orange tetrasodium salt
Xylenol orange tetrasodium salt
Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: ∼210 °C
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR