Alizarine

Supplier: Molekula
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2-Dihydroxyanthraquinone1,2-Dihydroxy-9,10-anthracenedione1,2-Dihydroxy-9,10-anthraquinoneAlizarin

MOLEM74253560EA 0 GBP
MOLEM74253560
Alizarine
Alizarine
Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Boiling Pt: 430 °C (1013 hPa)
Melting Pt: 290 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001201
CAS Number: 72-48-0
EINECS: 200-782-5
Merck Index: 13,00246

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR