2-Fluoro-6-(trifluoromethyl)benzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Cyano-3-fluorobenzotrifluoride

002047-1G 002047-5G 002047-25G
FLUO002047-1GEA 0 GBP
FLUO002047-1G FLUO002047-5G FLUO002047-25G
2-Fluoro-6-(trifluoromethyl)benzonitrile
2-Fluoro-6-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 26…28 °C
Density: 1.373 g/cm³ (20 °C)
MDL Number: MFCD00061153
CAS Number: 133116-83-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR