Diphenylamine 1% Reag. Ph. Eur. 1032102
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N-Phenylbenzeneamine
, N-Phenylaniline
87823.290EA
249
GBP
87823.290
Diphenylamine 1% Reag. Ph. Eur. 1032102
Diphenylamine
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formula:
(C₆H₅)₂NH MW: 169.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003014 CAS Number: 122-39-4 EINECS: 204-539-4 UN: 1830 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...