Methyl blue, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Disodium [[4-[bis[4-[(sulphonatophenyl)amino]phenyl]methylene]cyclohexa-2,5-dien-1-ylidene]amino]benzenesulphonatePoirriers blueAniline blue sodium saltCotton blueWater blueC.I. 42780Aniline blue, water soluble[[4-[bis[4-[(sulphophenyl)amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]amino]-benzenesulphonic acid sodium salt (1:2)Acid Blue 93

M-Clarity™ quality level = MQ200

M5528-25GEA 140 GBP
M5528-25G
Methyl blue, Sigma-Aldrich®
Methyl blue
Formula: C₃₇H₂₇N₃Na₂O₉S₃
MW: 799.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058509
CAS Number: 28983-56-4
EINECS: 249-352-9
Merck Index: 13,06055

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR