Zinc iodine chloride in aqueous solution Reag. Ph. Eur. 1096602
Supplier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
87726.180EA
231
GBP
87726.180
Zinc iodine chloride in aqueous solution Reag. Ph. Eur. 1096602
Zinc iodine chloride in aqueous solution
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Storage Temperature:
Ambient |
UN:
2920 ADR: 8 (3),II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...