Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution

Supplier: ASSOCXIATES OF CAPE COD INR. I
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Caustic soda

CAPEPH200EA 0 GBP
CAPEPH200
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR