Diethoxy(dimethyl)silane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Dimethyldiethoxysilane
M-Clarity™ quality level = MQ200
40120-100ML
40120-500ML
SIAL40120-100MLEA
85.8
GBP
SIAL40120-100ML
SIAL40120-500ML
Diethoxy(dimethyl)silane, Sigma-Aldrich®
Diethoxy(dimethyl)silane
Formula:
C₆H₁₆O₂Si MW: 148.28 g/mol Boiling Pt: 114 °C (1013 hPa) Melting Pt: –87 °C Density: 0.853 g/cm³ (20 °C) |
MDL Number:
MFCD00009068 CAS Number: 78-62-6 EINECS: 201-127-6 UN: 2380 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...