Pyridoxine hydrochloride

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Pyridoxine HCIPN HCl(5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloridePyridoxine HClPyridoxol hydrochlorideAdermine hydrochlorideVitamin B6 hydrochloride

47862.EA 33.1 GBP
47862.
Pyridoxine hydrochloride
Pyridoxine hydrochloride
Formula: C₈H₁₂ClNO₃
MW: 205.64 g/mol
Melting Pt: 214…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012807
CAS Number: 58-56-0
EINECS: 200-386-2
Merck Index: 13,08072
https://uk.vwr.com/stibo/web/std.lang.all/67/28/28506728.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR