2-Phenyl-1,3-propanediol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

SIAL559296-5GEA 192 GBP
SIAL559296-5G
2-Phenyl-1,3-propanediol, Sigma-Aldrich®
2-Phenyl-1,3-propanediol
Formula: HOH₂CCH(C₆H₅)CH₂OH
MW: 152.19 g/mol
MDL Number: MFCD00236056
CAS Number: 1570-95-2
EINECS: 411-810-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR