Disposable Wet Wipe
Supplier: Allied Hygiene Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LOCAB81080007EA 0 GBP
LOCAB81080007
Disposable Wet Wipe
Wipes Disposable Wipes and Towels Wipes for General Applications
A disposable wet wipe designed for the effective disinfection of all surfaces, preventing cross contamination in all healthcare areas.
  • Solution: 70% IPA
  • Odour: Slight pleasant
  • MSDS: 70% IPA Mix wipe
Tested and approved to European norms EN1276/ EN14348/EN13727 for bactericidal efficacy in medical environments and has effective kill rates against all potentially lethal micro-organisms including MRSA, HIV, C-Difficile, Enterococcus hirae. Medimax 70% IPA wipes have been tested and conform additionally to EN13624 fungicidal activity.
Also ideal for organic food production as the IPA wipe leaves no residue.