Potassium dihydrogen phosphate ≥99.00%, white crystals

Supplier: DISTRIBUTOR UK
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: prim.-Potassium phosphateMonopotassiumphosphatePotassium dihydrogen orthophosphatePotassium phosphate monobasicMonobasic Potassium PhosphateMonopotassium phosphate

DIUK10757214EA 97.1 GBP
DIUK10757214
Potassium dihydrogen phosphate ≥99.00%, white crystals
Potassium dihydrogen phosphate
Formula: KH₂PO₄
MW: 136.09 g/mol
Melting Pt: 253 °C
MDL Number: MFCD00011401
CAS Number: 7778-77-0
Merck Index: 15, 7780

Order Now

Specification Test Results

Melting Point 253 °C
Color White
pH 4.5
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99%
Merck Index 15, 7780
IUPAC Name potassium;dihydrogen phosphate
Formula Weight 136,09
Percent Purity ≥99%
Identification Pass Test
Loss on Drying 0,2% max.
Packaging Glass Bottle
Decomposition Information >250 °C
Chemical Name or Material Potassium Phosphate Monobasic

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR