Lead acetate cotton Reag. Ph. Eur. 1048101

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Lead di-acetateLead (II) acetate on cotton

85868.130EA 367 GBP
85868.130
Lead acetate cotton Reag. Ph. Eur. 1048101
Lead acetate cotton

  • Composition: 1 dilute acetic acid:10 lead acetate solution
  • Form: Cottony balls


The reagent is specially intended for use in testing Ph Eur and monographs. This solution was produced according 4.1.1 of Ph Eur specifications using Ph Eur reagent quality components and water meeting the Ph Eur monograph requirement for Purified Water.


Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: (H₃CCOO)₂Pb
MW: 325.29 g/mol
CAS Number: 6080-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR