Acetic anhydride 25% in pyridine Reag. Ph. Eur. 1000501

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Acetyl oxideAcetyl acetate

87760.290EA 988 GBP
87760.290
Acetic anhydride 25% in pyridine Reag. Ph. Eur. 1000501
Acetic anhydride

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: (CH₃CO)₂O
MW: 102.09 g/mol
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1282
ADR: 3,II
https://uk.vwr.com/stibo/web/std.lang.all/22/53/2302253.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR