Sodium azide
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Sodium triazide
EMR209050
EMR209100
EMR209500
EURLEMR209050EA
0
GBP
EURLEMR209050
EURLEMR209100
EURLEMR209500
Sodium azide
Sodium azide
Formula:
NaN₃ MW: 65.01 g/mol Melting Pt: 275 °C Density: 1.85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003536 CAS Number: 26628-22-8 EINECS: 247-852-1 UN: 1687 ADR: 6.1,II Merck Index: 13,08653 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...