X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
M-Clarity™ quality level = MQ100
703729
10703729001
ROCH703729EA
0
GBP
ROCH703729
ROCH10703729001
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula:
C₁₄H₁₅BrClNO₆ MW: 408.63 g/mol Melting Pt: 237…239 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005666 CAS Number: 7240-90-6 EINECS: 230-640-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...