Chromotropic acid disodium salt dihydrate, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate
, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate
, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate
, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate
M-Clarity™ quality level = MQ200
1.02498.0025EA
161
GBP
1.02498.9999
1.02498.0025
Chromotropic acid disodium salt dihydrate, Supelco®
Chromotropic acid disodium salt dihydrate
Formula:
(HO)₂C₁₀H₄(SO₃Na)₂·2H₂O MW: 400.29 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150612 CAS Number: 5808-22-0 EINECS: 204-972-9 Merck Index: 13,02261 |
Specification Test Results
Solubility | 170 g/l (20 °C) |
Melting Point |
|
Molar Mass | 400.30 g/mol |
Bulk Density | 780 kg/m3 |
pH | 3.6 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...