Chromotropic acid disodium salt dihydrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate
M-Clarity™ quality level = MQ200
27150-250G-F
27150-50G-F
SIAL27150-250G-FEA
993
GBP
SIAL27150-250G-F
SIAL27150-50G-F
Chromotropic acid disodium salt dihydrate, Sigma-Aldrich®
Chromotropic acid disodium salt dihydrate
Formula:
(HO)₂C₁₀H₄(SO₃Na)₂·2H₂O MW: 400.29 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150612 CAS Number: 5808-22-0 EINECS: 204-972-9 Merck Index: 13,02261 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...