Environmental Express® VOA Vials, Certified
Supplier: Cole Parmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Environmental Express®
COLE35204-91EA 117 GBP
COLE35204-91
Environmental Express® VOA Vials, Certified
Vials
Borosilicate glass vials meet EPA guidelines for contamination-free containers.
  • Select standard screw-top or open-top septa caps
  • Screw-top caps feature either PTFE or septa liner
Level 1 VOA vials meet EPA recommended guidelines for contamination-free sample containers and include a Certificate of Analysis. Septa offered as a liner option for closed caps to ensure greater seal integrity.