Extraction Unit for Ovens
Supplier: Genlab
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GENLOVENEXTEA 423 GBP
GENLOVENEXT
Extraction Unit for Ovens
Ovens Standard Ovens
This module sit on top of any standard Genlab oven and extracts air from the oven at double the normal extraction rate to increase drying time.
The enclosure is 200×200×150 mm with a 75 mm duct outlet and extraction is controlled from the front damper position. The unit can be turned on and off indpendently to the oven.