Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 6F7]

Supplier: Antibodies.com LLC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ANTIA85319-50EA 230 GBP
ANTIA85319-50 ANTIA85319-100
Anti-CRISPR-Cas9 Mouse Monoclonal Antibody [clone: 6F7]
Antibodies
Mouse monoclonal (6F7) antibody to CRISPR-Cas9 for WB, ICC/IF and IHC with samples derived from Staphylococcus aureus.

  • Validated applications: WB, ICC/IF, IHC
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:1000 to 1:500

Type: Primary
Antigen: CRISPR-Cas9
Clonality: Monoclonal
Clone: 6F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Staphylococcus aureus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR