Anti-ACOT1 Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ANTIA99842-100EA 325 GBP
ANTIA99842-100
Anti-ACOT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ACOT1 for WB, IF and ELISA with samples derived from human.

  • Validated applications: WB, IF, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:40000

Type: Primary
Antigen: ACOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR