Anti-ANKRD25 Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ANTIA99637-100EA 325 GBP
ANTIA99637-100
Anti-ANKRD25 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to KANK2 for IHC, IF and ELISA with samples derived from human.

  • Validated applications: IHC, IF, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:40000

Type: Primary
Antigen: ANKRD25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR