Anti-ABHD14B Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ANTIA98723-100EA 325 GBP
ANTIA98723-100
Anti-ABHD14B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ABHD14B for IHC and ELISA with samples derived from human and rat.

  • Validated applications: IHC, ELISA
  • Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:5000

Type: Primary
Antigen: ABHD14B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR