Anti-AAK1 Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ANTIA98307-100EA 325 GBP
ANTIA98307-100
Anti-AAK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to AAK1 for WB and ELISA with samples derived from human, mouse and rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000

Type: Primary
Antigen: AAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR