SealPlate® Breathable Sealing Film
Supplier: Elkay Plastics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SealPlate®
ELKASEA-LPLT-GXSEA 320 GBP
ELKASEA-LPLT-GXS
SealPlate® Breathable Sealing Film
Seals Microplate Seals
SealPlate breathable sealing film is a hydrophobic rayon film with end tabs, using medical grade adhesives.
  • Made from hydrophobic rayon
  • Full width end tabs
  • Pierceable
  • Recommended for gas exchange assays
It is recommended for cell and tissue culture applications where gas exchange is needed for growth. Elkay offers a broad range of sealing films, foils and mats for use in a range of applications.