Transwell® Polycarbonate Membrane Cell Culture Insert, Sterile

Supplier: Corning
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Transwell®
CORN3244EA 2350 GBP
CORN3244
Transwell® Polycarbonate Membrane Cell Culture Insert, Sterile
Inserts
Transwell cell culture inserts are convenient, easy-to-use permeable support devices for the study of both anchorage-dependent and anchorage-independent cell lines.

  • Treated for optimal cell attachment
  • Packaged 12 inserts in a 24 well plate, 4 plates per case
  • Membrane must be stained for cell visibility
  • Sterilized by gamma radiation
  • Round well shaped with flat bottom

Cell growth area: 0,33 cm²
Membrane pore size: 8 µm
Membrane diameter: 6,5 mm
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR