HPLC Columns, Hector M C18
Supplier: Hichrom Limited
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HISMC18-M31001539EA 636 GBP
HISMC18-M31001539
HPLC Columns, Hector M C18
Chromatography Columns
Hector M C18 is the most versatile and popular reversed phase. It is extremely suitable for the validation of various analytes. It has an about 17% carbon load and is fully end-capped. It provides high selectivity, efficiency, and a good peak shape.
  • Bonded phase: Octadecyl groups