Hexaamminecobalt trichloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride

M-Clarity™ quality level = MQ100

481521-100G 481521-25G
SIAL481521-100GEA 270 GBP
SIAL481521-100G SIAL481521-25G
Hexaamminecobalt trichloride, Sigma-Aldrich®
Hexaamminecobalt trichloride
Formula: H₁₈CoCl₃N₆
MW: 267.47 g/mol
Density: 1.702 g/cm³ (20 °C)
MDL Number: MFCD00036304
CAS Number: 10534-89-1
EINECS: 234-103-9
Merck Index: 12,04711

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR