Chlorpyrifosmethyl

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: O,O-Dimethyl O-3,5,6-trichloro-2-pyridyl phosphorothionateO,O-Dimethyl O-3,5,6-trichloro-2-pyridyl thiophosphate

EHERC11601000EA 194 GBP
EHERC11601000
Chlorpyrifosmethyl
Chlorpyrifos-methyl
Formula: C₇H₇Cl₃NO₃PS
MW: 322.54 g/mol
Melting Pt: 45.5…46.5 °C
CAS Number: 5598-13-0
EINECS: 227-011-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR