3-Thiopheneboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Thienylboronic acid, Thiophene-3-boronic acid

011010-5G 011010-25G
FLUO011010-5GEA 0 GBP
FLUO011010-5G FLUO011010-25G
3-Thiopheneboronic acid
3-Thiopheneboronic acid
Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 162…164 °C
MDL Number: MFCD00151851
CAS Number: 6165-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR