2-Thiopheneboronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Thiophen-2-ylboronic acid, Thiophene-2-boronic acid, 2-Thienylboronic acid

011009-5G 011009-25G
FLUO011009-5GEA 0 GBP
FLUO011009-5G FLUO011009-25G
2-Thiopheneboronic acid
2-Thiopheneboronic acid
Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 146…147 °C
MDL Number: MFCD00151850
CAS Number: 6165-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR